Learn More
Medchemexpress LLC Tannic acid | 1401-55-4 | 81.7% | 5 G
SDP
Catalog No. 5000409912
Encompass_Preferred
Tannic acid is a novel hERG channel blocker with an IC50 of 3.4 μM. It has been featured in over 5 cited publications and is associated with research in Potassium Channel, Inflammation/Immunology, and Cancer.
  • Acts as a novel hERG channel blocker.
  • Has an IC50 of 3.4 μM.
  • Applicable in research related to Potassium Channel.
  • Applicable in research related to Inflammation/Immunology.
  • Applicable in research related to Cancer.
Catalog No. 50-004-09912 Supplier Medchemexpress LLC Supplier No. HYB21365G
Tannic acid is a novel hERG channel blocker with an IC50 of 3.4 μM. It has been featured in over 5 cited publications and is associated with research in Potassium Channel, Inflammation/Immunology, and Cancer.
  • Acts as a novel hERG channel blocker.
  • Has an IC50 of 3.4 μM.
  • Applicable in research related to Potassium Channel.
  • Applicable in research related to Inflammation/Immunology.
  • Applicable in research related to Cancer.