Learn More
Medchemexpress LLC Tannic acid | 1401-55-4 | 81.7% | 500 MG
SDP
Catalog No. 5000410504
Encompass_Preferred
Tannic acid is a novel hERG channel blocker that significantly ameliorates phenotypes in AD skin lesions. It has been shown to reduce dermal changes in AD mice and increase PPARγ expression in AD skin sections. Additionally, tannic acid treatment suppresses the up-regulation of IL-1β, TNFα, TNFR1, and COX2 protein expressions observed in vehicle-treated AD mice.
  • Novel hERG channel blocker
  • Ameliorates phenotypes in AD skin lesions
  • Reduces dermal changes in AD mice
  • Increases PPARγ expression in AD skin sections
  • Suppresses up-regulation of inflammatory protein expressions
Catalog No. 50-004-10504 Supplier Medchemexpress LLC Supplier No. HYB2136500MG
Tannic acid is a novel hERG channel blocker that significantly ameliorates phenotypes in AD skin lesions. It has been shown to reduce dermal changes in AD mice and increase PPARγ expression in AD skin sections. Additionally, tannic acid treatment suppresses the up-regulation of IL-1β, TNFα, TNFR1, and COX2 protein expressions observed in vehicle-treated AD mice.
  • Novel hERG channel blocker
  • Ameliorates phenotypes in AD skin lesions
  • Reduces dermal changes in AD mice
  • Increases PPARγ expression in AD skin sections
  • Suppresses up-regulation of inflammatory protein expressions