Learn More
Medchemexpress LLC HY-B2136 1g Medchemexpress, Tannic acid CAS:1401-55-4 Purity:>98%
SDP
Catalog No. 501933077
Encompass_Preferred
Medchemexpress, HY-B2136 1g Tannic acid CAS:1401-55-4 Tannic acid is a novel hERG channel blocker with IC 50 of 3.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3077 Supplier Medchemexpress LLC Supplier No. HYB21361G
Medchemexpress, HY-B2136 1g Tannic acid CAS:1401-55-4 Tannic acid is a novel hERG channel blocker with IC 50 of 3.4 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.