Learn More
Alkali Scientific Transfer Pipette, 5mL, Individually Wrapped, Large Bulb, Graduated to 1mL, Sterile, 500/cs
SDP
Catalog No. 502335891
Encompass_Preferred
These unbreakable, one-piece dropping pipettes eliminate contaminated rubber bulbs, broken glass and exposure to infections materials. These disposable dropping pipettes are made of ultra-clear, low density polyethylene (LDPE) and are ideal for microbiology, blood banking, hematology, immunology, and urinalysis. sterile pipettes are excellent for tissue culture applications. Their low-affinity surface reduces sample, cell or protein loss. They can also be heat sealed for airtight sample storage. Their uniform drop size can be used for reproducible spotting of gels and TLC plates, and for dispensing into test plates, ID strips, PCR tubes or 96 well plates.
Catalog No. 50-233-5891 Supplier Alkali Scientific Supplier No. DP411311
These unbreakable, one-piece dropping pipettes eliminate contaminated rubber bulbs, broken glass and exposure to infections materials. These disposable dropping pipettes are made of ultra-clear, low density polyethylene (LDPE) and are ideal for microbiology, blood banking, hematology, immunology, and urinalysis. sterile pipettes are excellent for tissue culture applications. Their low-affinity surface reduces sample, cell or protein loss. They can also be heat sealed for airtight sample storage. Their uniform drop size can be used for reproducible spotting of gels and TLC plates, and for dispensing into test plates, ID strips, PCR tubes or 96 well plates.