Learn More
Apexbio Technology LLC TANSHINONE-I-20MG
SDP
Catalog No. 5000567413
Encompass_Preferred
Tanshinone I (CAS No 568-73-0) is a diterpenoid quinone derived from the roots of the Salvia miltiorrhiza plant widely used in traditional Chinese medicine It belongs to the category of natural product-based pharmacological research focusing on its anti-inflammatory and antitumor properties Mechanistically it acts by modulating pathways like NF- B and MAPK which play crucial roles in inflammation and cancer progression Tanshinone I has demonstrated significant in vitro activity often in the nanomolar to low micromolar range It is commonly used in cell models such as cancer cell lines and in animal studies to explore therapeutic potentials Moreover it serves in drug screening for novel anti-cancer or anti-inflammatory agents Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67413 Supplier Apexbio Technology LLC Supplier No. N184520
Tanshinone I (CAS No 568-73-0) is a diterpenoid quinone derived from the roots of the Salvia miltiorrhiza plant widely used in traditional Chinese medicine It belongs to the category of natural product-based pharmacological research focusing on its anti-inflammatory and antitumor properties Mechanistically it acts by modulating pathways like NF- B and MAPK which play crucial roles in inflammation and cancer progression Tanshinone I has demonstrated significant in vitro activity often in the nanomolar to low micromolar range It is commonly used in cell models such as cancer cell lines and in animal studies to explore therapeutic potentials Moreover it serves in drug screening for novel anti-cancer or anti-inflammatory agents Experimental concentrations typically depend on the specific research design