Learn More
TARGETMOL CHEMICALS INC Tanshinone I 25MG
SDP
Catalog No. 502879917
Encompass_Preferred
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tanshinone I (Tanshinone A), an active principle isolated from the herbal medicine Salvia miltiorrhiza, displays cytotoxicity against tumor cells. Purity 98.58%
Catalog No. 50-287-9917 Supplier TARGETMOL CHEMICALS INC Supplier No. T290725MG
Also available in 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Tanshinone I (Tanshinone A), an active principle isolated from the herbal medicine Salvia miltiorrhiza, displays cytotoxicity against tumor cells. Purity 98.58%