Learn More
Medchemexpress LLC Tanshinone I | 568-73-0 | 98.8% | 276.29 | 100 MG
SDP
Catalog No. 5000430287
Encompass_Preferred
Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 and rabbit recombinant cPLA2. It demonstrates anti-inflammatory activity in rat carrageenan-induced paw oedema and adjuvant-induced arthritis. It is for research use only.
  • Inhibits type IIA human recombinant sPLA2 and rabbit recombinant cPLA2
  • Shows anti-inflammatory activity in animal models
  • For research use only
Catalog No. 50-004-30287 Supplier Medchemexpress LLC Supplier No. HYN0134100MG
Tanshinone I is an inhibitor of type IIA human recombinant sPLA2 and rabbit recombinant cPLA2. It demonstrates anti-inflammatory activity in rat carrageenan-induced paw oedema and adjuvant-induced arthritis. It is for research use only.
  • Inhibits type IIA human recombinant sPLA2 and rabbit recombinant cPLA2
  • Shows anti-inflammatory activity in animal models
  • For research use only