Learn More
Medchemexpress LLC HY-N0134 5mg Medchemexpress, Tanshinone I CAS:568-73-0 Purity:>98%
SDP
Catalog No. 501933416
Encompass_Preferred
Medchemexpress, HY-N0134 5mg Tanshinone I CAS:568-73-0 Tanshinone I is an inhibitor of type IIA human recombinant sPLA 2 ( IC 50 =11 μM) and rabbit recombinant cPLA 2 ( IC 50 =82 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3416 Supplier Medchemexpress LLC Supplier No. HYN01345MG
Medchemexpress, HY-N0134 5mg Tanshinone I CAS:568-73-0 Tanshinone I is an inhibitor of type IIA human recombinant sPLA 2 ( IC 50 =11 μM) and rabbit recombinant cPLA 2 ( IC 50 =82 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.