Learn More
Enzo Life Sciences Tanshinone IIA (25mg). CAS: 568-72-9
SDP
Catalog No. 502010991
Encompass_Preferred
Tanshinone IIA is a diterpenoid naphthoquinone found in the traditional Chinese medicine Tanshen (Salvia sp). It inhibits AP-1 activity by suppressing jun-fos-DNA complex formation (IC50=0.22 µM) and induces apoptosis in a variety of cell lines Purity: ≥93% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (5mg/ml). Long Term Storage: +4°C.
Catalog No. 50-201-0991 Supplier Enzo Life Sciences Supplier No. BMLGR3360025
Tanshinone IIA is a diterpenoid naphthoquinone found in the traditional Chinese medicine Tanshen (Salvia sp). It inhibits AP-1 activity by suppressing jun-fos-DNA complex formation (IC50=0.22 µM) and induces apoptosis in a variety of cell lines Purity: ≥93% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (5mg/ml). Long Term Storage: +4°C.