Learn More
Apexbio Technology LLC Tanshinone IIA 568-72-9 20mg
SDP
Catalog No. 5000091727
Encompass_Preferred
Tanshinone IIA (CAS 568-72-9) is a diterpene quinone extracted from the medicinal plant Salvia miltiorrhiza (Danshen) It modulates cellular signaling by inhibiting the binding of transcription factors NF- B and AP-1 to DNA thereby regulating gene expression associated with inflammation and proliferation Tanshinone IIA has demonstrated the capacity to suppress -amyloid aggregation and protect neuronal PC12 cells against -amyloid-induced apoptosis It exhibits both antioxidant and anti-inflammatory properties supporting its use in studies related to oncology neurodegeneration and inflammation
Catalog No. 50-000-91727 Supplier Apexbio Technology LLC Supplier No. N184620
Tanshinone IIA (CAS 568-72-9) is a diterpene quinone extracted from the medicinal plant Salvia miltiorrhiza (Danshen) It modulates cellular signaling by inhibiting the binding of transcription factors NF- B and AP-1 to DNA thereby regulating gene expression associated with inflammation and proliferation Tanshinone IIA has demonstrated the capacity to suppress -amyloid aggregation and protect neuronal PC12 cells against -amyloid-induced apoptosis It exhibits both antioxidant and anti-inflammatory properties supporting its use in studies related to oncology neurodegeneration and inflammation