Learn More
Medchemexpress LLC Tanshinone IIA (Dan Shen ketone) | 568-72-9 | 99.78% | 294.34 | 5 MG
SDP
Catalog No. 5000430955
Encompass_Preferred
Tanshinone IIA is a primary component found in the root of Salvia miltiorrhiza Bunge. It may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2, and has anti-tumor effects including inhibiting tumor cell proliferation, disrupting the tumor cell cycle, promoting tumor cell apoptosis, and inhibiting tumor cell invasion and transfer.
  • Has antiproliferative activity against various cell lines
  • Protects rat myocardium-derived H9C2 cells against apoptosis
  • Blocks the PI3K/Akt/mTOR pathway in gastric carcinoma AGS cells
Catalog No. 50-004-30955 Supplier Medchemexpress LLC Supplier No. HYN01355MG
Tanshinone IIA is a primary component found in the root of Salvia miltiorrhiza Bunge. It may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2, and has anti-tumor effects including inhibiting tumor cell proliferation, disrupting the tumor cell cycle, promoting tumor cell apoptosis, and inhibiting tumor cell invasion and transfer.
  • Has antiproliferative activity against various cell lines
  • Protects rat myocardium-derived H9C2 cells against apoptosis
  • Blocks the PI3K/Akt/mTOR pathway in gastric carcinoma AGS cells