Learn More
TARGETMOL CHEMICALS INC Tanshinone IIA sulfonate sodiu
SDP
Catalog No. 502878874
Encompass_Preferred
Also available in 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Tanshinone IIA sulfonate sodium (Tanshinone IIA sodium sulfonate) is a water-soluble derivative of tanshinone IIA extracted from Savia miltiorrhiza; a potent negative allosteric modulator of the human purinergic receptor P2X7. Tanshinone IIA sulfonate sodium (12.5 uM) inhibits hypoxia-induced PKG and PPAR-gamma downregulation in PASMCs and distal pulmonary arteries of rats. Purity 99.9%
Catalog No. 50-287-8874 Supplier TARGETMOL CHEMICALS INC Supplier No. T2946100MG
Also available in 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Tanshinone IIA sulfonate sodium (Tanshinone IIA sodium sulfonate) is a water-soluble derivative of tanshinone IIA extracted from Savia miltiorrhiza; a potent negative allosteric modulator of the human purinergic receptor P2X7. Tanshinone IIA sulfonate sodium (12.5 uM) inhibits hypoxia-induced PKG and PPAR-gamma downregulation in PASMCs and distal pulmonary arteries of rats. Purity 99.9%