Learn More
Medchemexpress LLC Tanshinone IIA sulfonate sodium | 69659-80-9 | 396.39 | 5 MG
SDP
Catalog No. 5000414800
Encompass_Preferred
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA. It acts as an inhibitor of store-operated Ca2+ entry (SOCE) and is used to treat cardiovascular disorders. This product is for research use only and not sold to patients.
  • Inhibits store-operated Ca2+ entry (SOCE)
  • Used to treat cardiovascular disorders
  • Inhibits hypoxia-induced PKG and PPAR-γ downregulation
  • Suppresses TRPC1 and TRPC6 expression in hypoxic PASMCs
  • Inhibits CYP3A4 activity
  • Attenuates scopolamine-induced cognitive dysfunctions
  • Alleviates hypoxia-induced characteristic changes in chronic hypoxia PH rat model
  • Prevents peritoneal adhesion
Catalog No. 50-004-14800 Supplier Medchemexpress LLC Supplier No. HYN13705MG
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA. It acts as an inhibitor of store-operated Ca2+ entry (SOCE) and is used to treat cardiovascular disorders. This product is for research use only and not sold to patients.
  • Inhibits store-operated Ca2+ entry (SOCE)
  • Used to treat cardiovascular disorders
  • Inhibits hypoxia-induced PKG and PPAR-γ downregulation
  • Suppresses TRPC1 and TRPC6 expression in hypoxic PASMCs
  • Inhibits CYP3A4 activity
  • Attenuates scopolamine-induced cognitive dysfunctions
  • Alleviates hypoxia-induced characteristic changes in chronic hypoxia PH rat model
  • Prevents peritoneal adhesion