Learn More
Medchemexpress LLC Tanshinone IIA sulfonate sodium | 69659-80-9 | 396.39 | 500 MG
SDP
Catalog No. 5000414724
Encompass_Preferred
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA. This compound acts as an inhibitor of store-operated Ca2+ entry (SOCE) and is primarily used in the treatment of cardiovascular disorders. It is intended for research purposes only.
  • Inhibits hypoxia-induced PKG and PPAR-γ downregulation in PASMCs.
  • Suppresses TRPC1 and TRPC6 expression.
  • Inhibits basal calcium concentration and SOCE.
  • Primarily inhibits CYP3A4 activities in vitro.
  • Shows protective effects against Aβ-induced cell toxicity in SH-SY5Y cells.
  • Alleviates hypoxia-induced characteristic changes in chronic hypoxia PH rat models.
  • Effectively prevents peritoneal adhesion in rats.
Catalog No. 50-004-14724 Supplier Medchemexpress LLC Supplier No. HYN1370500MG
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA. This compound acts as an inhibitor of store-operated Ca2+ entry (SOCE) and is primarily used in the treatment of cardiovascular disorders. It is intended for research purposes only.
  • Inhibits hypoxia-induced PKG and PPAR-γ downregulation in PASMCs.
  • Suppresses TRPC1 and TRPC6 expression.
  • Inhibits basal calcium concentration and SOCE.
  • Primarily inhibits CYP3A4 activities in vitro.
  • Shows protective effects against Aβ-induced cell toxicity in SH-SY5Y cells.
  • Alleviates hypoxia-induced characteristic changes in chronic hypoxia PH rat models.
  • Effectively prevents peritoneal adhesion in rats.