Learn More
Medchemexpress LLC Tanshinone IIA sulfonate sodium | 69659-80-9 | MFCD09028094 | 396.39 | 50 MG
SDP
Catalog No. 5000415796
Encompass_Preferred
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, acting as an inhibitor of store-operated Ca2+ entry (SOCE), and is utilized in the treatment of cardiovascular disorders. It is intended for research use only and is not sold to patients.
  • Acts as an inhibitor of store-operated Ca2+ entry (SOCE).
  • Used to treat cardiovascular disorders.
  • Inhibits hypoxia-induced PKG and PPAR-γ downregulation.
  • Suppresses TRPC1 and TRPC6 expression and reverses suppressive effects on basal calcium concentration and SOCE in hypoxic PASMCs.
  • Inhibits CYP3A4 activity.
  • Attenuates scopolamine-induced cognitive dysfunctions and improves the cholinergic system.
  • Increases superoxide dismutase activity and decreases oxidative stress markers.
  • Alleviates hypoxia-induced changes in chronic hypoxia PH rat model.
  • Prevents peritoneal adhesion.
Catalog No. 50-004-15796 Supplier Medchemexpress LLC Supplier No. HYN137050MG
Tanshinone IIA sulfonate (sodium) is a derivative of tanshinone IIA, acting as an inhibitor of store-operated Ca2+ entry (SOCE), and is utilized in the treatment of cardiovascular disorders. It is intended for research use only and is not sold to patients.
  • Acts as an inhibitor of store-operated Ca2+ entry (SOCE).
  • Used to treat cardiovascular disorders.
  • Inhibits hypoxia-induced PKG and PPAR-γ downregulation.
  • Suppresses TRPC1 and TRPC6 expression and reverses suppressive effects on basal calcium concentration and SOCE in hypoxic PASMCs.
  • Inhibits CYP3A4 activity.
  • Attenuates scopolamine-induced cognitive dysfunctions and improves the cholinergic system.
  • Increases superoxide dismutase activity and decreases oxidative stress markers.
  • Alleviates hypoxia-induced changes in chronic hypoxia PH rat model.
  • Prevents peritoneal adhesion.