Learn More
Selleck Chemical LLC Tanshinone IIA 50mg 568-72-9 Tanshinone B
SDP
Catalog No. 505446
Encompass_Preferred
Tanshinone IIA (Tanshinone B) is the most abundant diterpene quinone in Danshen, Salviae miltiorrhizae Radix, a widely prescribed traditional herbal medicine that is used to treat cardiovascular and inflammatory diseases; A natural monoacylglycerol lipase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-544-6 Supplier Selleck Chemical LLC Supplier No. S236550MG
Tanshinone IIA (Tanshinone B) is the most abundant diterpene quinone in Danshen, Salviae miltiorrhizae Radix, a widely prescribed traditional herbal medicine that is used to treat cardiovascular and inflammatory diseases; A natural monoacylglycerol lipase inhibitor. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.