Learn More
Alkali Scientific SureSeal™ Sterile Transport / Mailing Tubes, 5mL with Attached Screw Cap, 100 per Sterile Bag, 1000/cs
SDP
Supplier:  Alkali Scientific C3581
Encompass_Preferred
Transport tubes, sometimes called mailing tubes, are used in multiple sectors of the laboratory market, including medical, hospital, environmental, independent testing, food testing, and biological research labs. Used to store, transport or freeze samples, these tubes come in 5mL (16 x 60mm) and are fitted with leak-proof screw-caps. They are made of high-grade polypropylene, and include white printed graduations and marking area. Tubes are self-standing and come packaged in 10 bags 100 tubes.
Catalog No. 50-233-6022