Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tantalum

Catalog No. 357243225CM Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 cm2
225 cm2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
35-724-3225CM 225 cm2
35-724-3100CM 100 cm2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 35-724-3225CM Supplier Sigma Aldrich Supplier No. 357243225CM2
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tantalum

Specifications

Melting Point 2996°C
CAS 7440-25-7
Boiling Point 5425°C
Density 16.69 g/cm3 (literature)
Quantity 225 cm2
Molecular Formula Ta
Linear Formula Ta
MDL Number MFCD00011252
Synonym Ta
Molecular Weight (g/mol) 180.95
Percent Purity ≥99.9% trace metals basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.