Learn More
Electron Microscopy Sciences Tantalum Boat
SDP
Catalog No. 5027131
Encompass_Preferred
For disc aperture cleaning, vacuum evaporation. Thickness: 0.05 mm (0.002") Overall Length is 75 mm (3"). Trough Measures: 12 mm(1.2")long x 5 mm (3/16")wide x 1 mm deep
Catalog No. 50-271-31 Supplier Electron Microscopy Sciences Supplier No. 73814
For disc aperture cleaning, vacuum evaporation. Thickness: 0.05 mm (0.002") Overall Length is 75 mm (3"). Trough Measures: 12 mm(1.2")long x 5 mm (3/16")wide x 1 mm deep