Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum carbide, 99.5% (metals basis)

Catalog No. AA1214430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1214430 250 g
AA1214409 10 g
AA1214418 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1214430 Supplier Thermo Scientific Chemicals Supplier No. 01214430
Add to Cart

CAS: 12070-06-3 | CTa | 192.96 g/mol

Tantalum carbides is used in  tungsten carbide. It is also used as a coating for steel moulds in the injection moulding of aluminum alloys. While providing a hard, wear resistant surface, it also provides a low friction mould surface. It is also used in the production of sharp instruments with extreme mechanical resistance and hardness, tool bits for cutting tools.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12070-06-3
Molecular Formula CTa
Molecular Weight (g/mol) 192.96
MDL Number MFCD00011255
InChI Key DUMHRFXBHXIRTD-UHFFFAOYSA-N
IUPAC Name methanidylidynetantalumylium
SMILES [C-]#[Ta+]

Specifications

Boiling Point 5500°C
Melting Point 3880°C
Color Black to Gray
Physical Form Powder
Quantity 250 g
UN Number UN3178
Solubility Information Soluble in hydrofluoric acid and nitric acid. Partially soluble in water.
Formula Weight 192.96
Percent Purity 99.5%
Odor Odorless
Density 13.9 g/mL
Assay (metals basis)
Chemical Name or Material Tantalum carbide
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H228
Flammable solid.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
EINECSNumber 235-118-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.