Alfa Aesar™ Tantalum nitride, 99.5% (metals basis)

Manufacturer: Alfa Aesar™ 1309318

Quantity 50g

 View more versions of this product

Catalog No. AA1309318

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 12033-62-4
    Formula Weight 194.95
    MDL Number MFCD00049568
    Chemical Name or Material Tantalum nitride

    Specifications

    Formula Weight 194.95
    TSCA TSCA
    Odor Odorless
    MDL Number MFCD00049568
    CAS 12033-62-4
    Mesh Size −325 Mesh
    Physical Form Powder
    Melting Point 3360°C
    Quantity 50g
    Chemical Name or Material Tantalum nitride
    Assay 99.5%, (metals basis)

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit