Learn More
Perkin Elmer US LLC Tantalum (Ta) Pure Standard, 10,000 µg/mL, H2O/8% HF, 500 mL
Catalog No. 502111850
Encompass_Preferred
Concentration of Element includes: Element Concentration Sc 100 µg/mL Ge 50 µg/mL Rn, ln, Tb 1 µg/mL Matrix 2 % HNO3+/- 0.5%
Catalog No. 50-211-1850 Supplier Perkin Elmer US LLC Supplier No. N9304342
Concentration of Element includes: Element Concentration Sc 100 µg/mL Ge 50 µg/mL Rn, ln, Tb 1 µg/mL Matrix 2 % HNO3+/- 0.5%