Learn More
Strem, An Ascensus Company CAS# 7721-01-9. 10g. Tantalum(V) chloride, anhydrous (99.9%-Ta). MFCD00011253
SDP
Catalog No. 5090112994
Encompass
CAS# 7721-01-9. 10g. Tantalum(V) chloride, anhydrous (99.9%-Ta). MFCD00011253. Molecular Weight: 358.21. Molecular Formula: TaCl5. Color/form: white to yellow pwdr. Strem# 73-1000. http://www.strem.com/catalog/v/73-1000/
Catalog No. 50-901-12994 Supplier Strem, An Ascensus Company Supplier No. 73100010G
CAS# 7721-01-9. 10g. Tantalum(V) chloride, anhydrous (99.9%-Ta). MFCD00011253. Molecular Weight: 358.21. Molecular Formula: TaCl5. Color/form: white to yellow pwdr. Strem# 73-1000. http://www.strem.com/catalog/v/73-1000/