Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum(V) chloride, anhydrous, Puratronic™, 99.995% (metals basis)

Catalog No. AA4373106
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4373106 5 g
AA4373114 25 g
AA4373122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4373106 Supplier Thermo Scientific Chemicals Supplier No. 04373106
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7721-01-9 | Cl5Ta | 358.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7721-01-9
Molecular Formula Cl5Ta
Molecular Weight (g/mol) 358.20
MDL Number MFCD00011253
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
Synonym Tantalum pentachloride
IUPAC Name tantalum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]

Specifications

Boiling Point 242°C
Melting Point 216°C
Color White to Yellow
Physical Form Powder
Quantity 5 g
Odor Acidic
Packaging Ampule
Sensitivity Moisture sensitive
Density 3.68 g/mL
UN Number UN3260
Merck Index 14,9054
Formula Weight 358.21
Percent Purity 99.995%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Tantalum (V) chloride, Anhydrous
Show More Show Less
Hazard Category H302-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 231-755-6
RTECSNumber WW5600000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.