Learn More
Tantalum(V) chloride, Puratronic™, 99.99% (metals basis)
Description
Tantalum(V) chloride is used in the preparation of catalyst for the polycyclotrimerizations of alkenediynes, chloro-aryloxide compounds and for the plasma-enhanced atomic layer deposition of tantalum nitride films. It acts as a starting material for new edge-bridged octahedral M6 cluster compounds. It is involved in the preparation of tantalum(V) oxychloride, tantalum pentoxide. It is electrophilic and behaves like a Friedel-Crafts type catalyst, similar to aluminum(III) chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Boiling Point | 242°C |
| Melting Point | 216°C |
| Color | White to Yellow |
| Physical Form | Powder |
| Quantity | 25 g |
| UN Number | UN3260 |
| Merck Index | 14,9054 |
| Solubility Information | Soluble in ether,carbon tetrachloride,sulfuric acid and potassium hydroxide. Slightly soluble in ethanol. |
| Formula Weight | 358.21 |
| Percent Purity | 99.99% |
| Show More |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: II |
| EINECSNumber | 231-755-6 |
| RTECSNumber | WW5600000 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.