Learn More
Strem, An Ascensus Company CAS# 6074-84-6. 5 x 50g. Tantalum(V) ethoxide (99.99+%-Ta) PURATREM. MFCD00049785
SDP
Catalog No. 5090116269
Encompass
CAS# 6074-84-6. 5 x 50g. Tantalum(V) ethoxide (99.99+%-Ta) PURATREM. MFCD00049785. Molecular Weight: 406.26. Molecular Formula: Ta(OC2H5)5. Color/form: colorless to yellow liq. Strem# 93-7303. http://www.strem.com/catalog/v/93-7303/
Catalog No. 50-901-16269 Supplier Strem, An Ascensus Company Supplier No. 937303250G
CAS# 6074-84-6. 5 x 50g. Tantalum(V) ethoxide (99.99+%-Ta) PURATREM. MFCD00049785. Molecular Weight: 406.26. Molecular Formula: Ta(OC2H5)5. Color/form: colorless to yellow liq. Strem# 93-7303. http://www.strem.com/catalog/v/93-7303/