Tantalum(V) Fluoride, 99.9% (Metal Basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1405209

Quantity 10g

 View more versions of this product

Catalog No. AA1405209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7783-71-3
    Physical Form Powder
    Assay Percent Range 99.9%, metal basis
    Molecular Formula TaF5
    Formula Weight 275.94
    MDL Number MFCD00042542
    Synonym Tantalum pentafluoride
    Chemical Name or Material Tantalum(V) Fluoride

    Specifications

    Formula Weight 275.94
    Quantity 10g
    DOT Information DOT Class: 8
    EINECS Number 232-022-3
    Physical Form Powder
    Decomposition Information Decomposes in water forming tantalum oxyfluoride
    Density 4.74
    Assay Percent Range 99.9%, metal basis
    Molecular Formula TaF5
    CAS 7783-71-3
    Sensitivity Moisture sensitive
    MDL Number MFCD00042542
    Synonym Tantalum pentafluoride
    Health Hazard 1 Harmful by inhalation, in contact with skin and if swallowed. Causes burns.
    Chemical Name or Material Tantalum(V) Fluoride

    Harmful by inhalation, in contact with skin and if swallowed. Causes burns.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit