Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tantalum(V) oxide

Catalog No. 30351825G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-351-825G 25 g
30-351-8100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-351-825G Supplier Sigma Aldrich Supplier No. 30351825G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tantalum pentoxide is dielectric in nature making it useful for making capacitors in the electronics industry.Tantalum oxide is intermixed with silicon in gate dielectric structures. It may be used to synthesize hexagonally packed mesoporous tantalum oxide molecular sieves.Ion-beam sputter deposition of tantalum oxide films was investigated for possible optical coating applications.It may be used in making optical glass for lenses and in electronic circuits.

Specifications

CAS 1314-61-0
Quantity 25 g
Molecular Formula O5Ta2
Linear Formula Ta2O5
MDL Number MFCD00011254
Synonym Tantalum pentoxide
Molecular Weight (g/mol) 441.89
Percent Purity 99% trace metals basis
Density 8.2 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.