Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tantalum(V) oxide, 99.95% (metals basis excluding Nb), Nb <200ppm

Catalog No. AA3634322
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3634322 100 g
AA3634314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3634322 Supplier Thermo Scientific Chemicals Supplier No. 03634322
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1314-61-0 | O5Ta2 | 441.89 g/mol

Tantalum (V) oxide (Ta2O5) has potential applications as part of an environmental barrier coating system for Si3N4-based turbine components. Octadecyl phosphoric acid ester has been found to produce oriented, well-ordered monolayers on a flat tantalum (V) oxide surface, via self-assembly from a heptane/propan-2-ol solution. Tantalum(V) oxide (Ta 2 O 5 ) has been used extensively as an insulating layer in discrete capacitors and shows a high potential for semiconductor applications such as an insulating dielectric in the capacitive element in memory devices such as dynamic random access.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-61-0
Molecular Formula O5Ta2
Molecular Weight (g/mol) 441.89
MDL Number MFCD00011254
InChI Key PBCFLUZVCVVTBY-UHFFFAOYSA-N
IUPAC Name [(dioxotantalio)oxy]dioxotantalum
SMILES O=[Ta](=O)O[Ta](=O)=O

Specifications

Melting Point 1800°C
Physical Form Sintered Lump
Quantity 100 g
Odor Odorless
Density 8.2 g/mL
Merck Index 14,9056
Solubility Information Insoluble in water.
Formula Weight 441.89
Percent Purity 99.95%
Assay (metals basis excluding Nb), Nb <200ppm
Chemical Name or Material Tantalum(V) oxide
Show More Show Less
Hazard Category H500
EINECSNumber 215-238-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.