Learn More
Strem, An Ascensus Company CAS# 7440-25-7. 1m. Tantalum wire (99.95%). MFCD00011252
SDP
Catalog No. 5090112981
Encompass
CAS# 7440-25-7. 1m. Tantalum wire (99.95%). MFCD00011252. Molecular Weight: 180.95. Molecular Formula: Ta. Color/form: 0.5mm dia. (~3.3g/m) Strem# 73-0080. http://www.strem.com/catalog/v/73-0080/
Catalog No. 50-901-12981 Supplier Strem, An Ascensus Company Supplier No. 7300801MT
CAS# 7440-25-7. 1m. Tantalum wire (99.95%). MFCD00011252. Molecular Weight: 180.95. Molecular Formula: Ta. Color/form: 0.5mm dia. (~3.3g/m) Strem# 73-0080. http://www.strem.com/catalog/v/73-0080/