Learn More
Medchemexpress LLC Tanuxiciclib trihydrochloride | 1983984-11-7 | ≥98.0% | 588.94 g/mol | C26H28Cl3N5O3 | 5 MG
SDP
Catalog No. 5000221473
Encompass_Preferred
Tanuxiciclib trihydrochloride is the trihydrochloride salt of tanuxiciclib, a cyclin-dependent kinase (CDK) inhibitor supplied as a research-grade solid for biochemical and cell-based studies of CDK signaling and cell-cycle regulation. The compound is provided with high chemical purity and is intended for laboratory research applications only.
Catalog No. 50-002-21473 Supplier Medchemexpress LLC Supplier No. HY145599A5MG
Tanuxiciclib trihydrochloride is the trihydrochloride salt of tanuxiciclib, a cyclin-dependent kinase (CDK) inhibitor supplied as a research-grade solid for biochemical and cell-based studies of CDK signaling and cell-cycle regulation. The compound is provided with high chemical purity and is intended for laboratory research applications only.