Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tanuxiciclib (trihydrochloride) | 1983984-11-7 | 98.0% | C15H16Cl3FN6O | 1 MG
SDP

Catalog No. 5000354116
Encompass_Preferred

Tanuxiciclib trihydrochloride is a cyclin dependent kinase (CDK) inhibitor. This compound is provided as an off-white to light yellow solid with a purity of 98.0%. It is suitable for use in laboratory chemical applications and the manufacture of substances, particularly in cancer research targeting cell cycle regulation and metabolism.

  • Acts as a cyclin dependent kinase (CDK) inhibitor
  • Solid appearance
  • Off-white to light yellow color
  • Purity of 98.0%
  • Molecular weight: 421.68
  • Recommended storage at 4°C, sealed, away from moisture
  • Soluble in DMSO for in vitro studies
  • Not classified as a hazardous substance or mixture

Catalog No. 50-003-54116 Supplier Medchemexpress LLC Supplier No. HY145599A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tanuxiciclib trihydrochloride is a cyclin dependent kinase (CDK) inhibitor. This compound is provided as an off-white to light yellow solid with a purity of 98.0%. It is suitable for use in laboratory chemical applications and the manufacture of substances, particularly in cancer research targeting cell cycle regulation and metabolism.

  • Acts as a cyclin dependent kinase (CDK) inhibitor
  • Solid appearance
  • Off-white to light yellow color
  • Purity of 98.0%
  • Molecular weight: 421.68
  • Recommended storage at 4°C, sealed, away from moisture
  • Soluble in DMSO for in vitro studies
  • Not classified as a hazardous substance or mixture

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.