Learn More
Selleck Chemical LLC Tanzisertib HCl
SDP
Catalog No. 5000001980
Encompass_Preferred
Tanzisertib HCl(CC-930 JNK-930 JNKI-1) is kinetically competitive with ATP in the JNK-dependent phosphorylation of the protein substrate c-Jun and potent against all isoforms of JNK (Ki(JNK1) 44 3 nM IC50(JNK1) 61 nM Ki(JNK2) 6 2 0 6 nM IC50(JNK2) 5 nM IC50(JNK3) 5 nM) and selective against MAP kinases ERK1 and p38a with IC50 of 0 48 and 3 4 M respectively
Catalog No. 50-000-01980 Supplier Selleck Chemical LLC Supplier No. S84901G
Add to Cart
Add to Cart
Tanzisertib HCl(CC-930 JNK-930 JNKI-1) is kinetically competitive with ATP in the JNK-dependent phosphorylation of the protein substrate c-Jun and potent against all isoforms of JNK (Ki(JNK1) 44 3 nM IC50(JNK1) 61 nM Ki(JNK2) 6 2 0 6 nM IC50(JNK2) 5 nM IC50(JNK3) 5 nM) and selective against MAP kinases ERK1 and p38a with IC50 of 0 48 and 3 4 M respectively