Learn More
Medchemexpress LLC HY-112136 5mg Medchemexpress, TAO Kinase inhibitor 1 CAS:850467-66-2 Purity:>98%
SDP
Catalog No. 501969036
Encompass_Preferred
Medchemexpress, HY-112136 5mg TAO Kinase inhibitor 1 CAS:850467-66-2 TAO Kinase inhibitor 1 (compound 43) is a selective, ATP-competitive thousand-and-one amino acid kinases (TAOK) inhibitor with IC50s of 11 to 15 nM for TAOK1 and 2, respectively. TAO Kinase inhibitor 1 delays mitosis and induces mitotic cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9036 Supplier Medchemexpress LLC Supplier No. HY1121365MG
Medchemexpress, HY-112136 5mg TAO Kinase inhibitor 1 CAS:850467-66-2 TAO Kinase inhibitor 1 (compound 43) is a selective, ATP-competitive thousand-and-one amino acid kinases (TAOK) inhibitor with IC50s of 11 to 15 nM for TAOK1 and 2, respectively. TAO Kinase inhibitor 1 delays mitosis and induces mitotic cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.