Learn More
Medchemexpress LLC TAO Kinase inhibitor 1 | 850467-66-2 | 99.8% | 384.47 | 100 MG
SDP
Catalog No. 5000436169
Encompass_Preferred
TAO Kinase inhibitor 1 (compound 43) is a selective, ATP-competitive thousand-and-one amino acid kinases (TAOK) inhibitor with IC50s of 11 to 15 nM for TAOK1 and 2. It delays mitosis and induces mitotic cell death. It is for research use only.
  • Selective, ATP-competitive TAOK inhibitor.
  • IC50s of 11 to 15 nM for TAOK1 and 2.
  • Delays mitosis.
  • Induces mitotic cell death.
  • Purity: 99.81%.
Catalog No. 50-004-36169 Supplier Medchemexpress LLC Supplier No. HY112136100MG
TAO Kinase inhibitor 1 (compound 43) is a selective, ATP-competitive thousand-and-one amino acid kinases (TAOK) inhibitor with IC50s of 11 to 15 nM for TAOK1 and 2. It delays mitosis and induces mitotic cell death. It is for research use only.
  • Selective, ATP-competitive TAOK inhibitor.
  • IC50s of 11 to 15 nM for TAOK1 and 2.
  • Delays mitosis.
  • Induces mitotic cell death.
  • Purity: 99.81%.