Learn More
TARGETMOL CHEMICALS INC TAO KINASE INHIBITOR 2 5MG
SDP
Catalog No. 502579143
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. TAO Kinase inhibitor 2 is a TAO kinase inhibitor with an IC50 of 50-500 nM. TAO Kinase inhibitor 2 inhibits KIAA1361 and JIK with IC50s of 50-500 nM. purity: 98%
Catalog No. 50-257-9143 Supplier TARGETMOL CHEMICALS INC Supplier No. T775015MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. TAO Kinase inhibitor 2 is a TAO kinase inhibitor with an IC50 of 50-500 nM. TAO Kinase inhibitor 2 inhibits KIAA1361 and JIK with IC50s of 50-500 nM. purity: 98%