Learn More
Eurofins Discoverx TAO1, active, 250 µg. (14-749M)
Supplier:  Eurofins Discoverx 14749M
Encompass
N-terminal 6xHis-tagged recombinant, human TAO1 amino acids 1-327. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9284