Learn More
Eurofins Discoverx TAO2, active, 10 µg. (14-736)
Supplier:  Eurofins Discoverx 14736
Encompass
N-terminal 6xHis-tagged recombinant, TAO2. TAO2 is a mitogen-activated protein kinase kinase kinase (MAP3K) that doubly phosphorylates and activates the MAP kinase kinases (MAP2Ks) MEK3 and MEK6. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9261