Learn More
Eurofins Discoverx TAO3, active, 250 µg. (14-745M)
Supplier:  Eurofins Discoverx 14745M
Encompass
N-terminal 6xHis-tagged recombinant, human TAO3 amino acids 1-411; N47S. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9278