Learn More
Sino Biological Anti-TAOK3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201029T08100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human TAOK3 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-224-7670