Learn More
Gibco™ TAP Growth Media, optimized for Chlamydomonas culture

Description
Gibco™ TAP Media is optimized for the growth and maintenance of Chlamydomonas reinhardtii. The formulation comes ready-to-use at 1X to let you avoid laborious media preparation steps.
Each Gibco™ TAP Media comes in the award winning Gibco™ bottle designed for easier use in the biosafety cabinet, minimizing the risk of contamination, and helping you perform cell culture more consistently. Together, the superior packaging and quality, greater reliability, and improved consistency in Chlamydomonas culture, results in better overall efficiency and more robust data (Fig. 1, 2).
Specifications
Specifications
| Form | Liquid |
| Product Type | Growth Medium |
| Quantity | 1 L |
| Shipping Condition | Room Temperature |
| Preparation Method | None (Ready-to-Use) |
| Species | C. reinhardtii |
| Content And Storage | Gibco™ TAP Growth Media contains: • 1 L Gibco™ TAP Media — Store at 4 °C |
Frequently Asked Questions (FAQs)
Please see the following suggestions:
1. For best results, the algae need to be between 1 x 10e6 and 2 x 10e6 cells/mL (thus, an OD of 0.3-0.5). The concentration of the cells should not exceed 3 x 106 cells/mL. Cell growth can be measured by OD750 and the linear range will be between 0.2 and 1.2 (in 1 cm lightpass). If the OD is out of the linear range, please dilute the cells and measure again to get an accurate reading.
The formula is: cell number = (OD750 - 0.088)/9 million/mL
2. Transformation of linearized DNA is much more efficient (~70% more efficient) than transformation of circular DNA.
3. The quality and concentration of the DNA are critical to the transformation efficiency. You will want to use PureLink HQ, PureLink HiPure, or equivalent plasmid purification kits for pure DNA. It is better to have a small volume of concentrated pure DNA than a large volume of DNA of low concentration. (If you are transforming linearized plasmid, then after the linearization you will need to clean the plasmid up using either gel extraction or a PCR cleanup kit prior to transformation.) We recommend using 2 µg of linearized plasmid DNA per electroporation.
4. Insertion of the plasmid DNA into the genome occurs randomly. On average, only 20% of transformants will express the gene of interest at appreciable levels. We recommend first screening the colonies by colony PCR to ensure full integration of the promoter and gene of interest, followed by the screening of several positive clones for the expression of the gene of interest to pick the highest expressing clone.
5. Because the C. reinhardtii genome has a very high GC content (~62% GC), the expression levels of recombinant genes are significantly improved if the gene of interest is adapted to the preferred codon usage of highly expressed C. reinhardtii genes.
6. Since the transformation efficiency depends on the random integration of the construct into the genome, the results of the electroporation will depend on the nature of the gene of interest. You can try to transform the control vector that comes with the kit to confirm that the kit and their electroporation method are working correctly.
Unfortunately, silencing is a big problem in algae. The extent of silencing depends on the sequence of the gene and the toxicity of the protein that expresses from that gene to the cell. Our pChlamy_4 vector was designed in order to circumvent the transgene silencing that often occurs in C. reinhardtii. This vector is also designed so that proteins are expressed as transcriptional fusions with the blemoycin/zeocin resistance gene (sh-ble). The self-cleaving sequence for the 2A peptide from the foot-and-mouth-disease-virus (FMDV) is placed between the antibiotic resistance gene and the gene of interest. It encodes a short ~20 amino acid sequence that mediates proper cleavage to yield two discrete proteins. With this system we have seen positive transformants maintain high expression levels for much longer than with other systems, even after many passages with or without selection pressure.
The pChlamy_4 vector was designed in order to circumvent the transgene silencing that often occurs in C. reinhardtii. This vector is also designed so that proteins are expressed as transcriptional fusions with the blemoycin/zeocin resistance gene (sh-ble). The self-cleaving sequence for the 2A peptide from the foot-and-mouth-disease-virus (FMDV) is placed between the antibiotic resistance gene and the gene of interest. It encodes a short ~20 amino acid sequence that mediates proper cleavage to yield two discrete proteins. With this system we have seen positive transformants maintain high expression levels for much longer than with other systems, even after many passages with or without selection pressure.
The pChlamy_1 vector does not have the stop codon and the 3' UTR. The pChlamy_3 vector has the 3'UTR that has been shown to increase protein expression.
Yes. For the TOPO version, you can design the primer to make sure the coding sequence is in frame with the ATG in the vector.
For Research Use Only. Not intended for human or animal therapeutic or diagnostic use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.