Learn More
YourBio Health TAP MicroSelect Pain Free Blood Collection Device
Catalog No. 21010299
Change view
Click to view available options
Capacity (Metric):
500 μL
Catalog No. Capacity (Metric)
21-010-299 500 μL
1 options
Catalog No. 21-010-299 Supplier YourBio Health Supplier No. 770100
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Designed to deliver virtually painless, large volume collection of up to 500 μ of whole with capillary high-quality blood
  • Push-button, virtually painless capillary blood collection enabled by HALO microneedle array technology
  • Collects high-quality whole capillary blood up to 900 μL per sample for a broad range of standard tests
  • Single-use, self-contained device designed for simple upper-arm application and easy disposal
  • FDA 510 (k) cleared in the U.S. and CE marked in the EU
  • Ideal for decentralized clinical trials, telehealth programs, at-home wellness screening, and clinic workflows reduces reliance on venipuncture and fingersticks
  • Demonstrated strong user experience
  • Compact and lightweight (~30 g) with a 24 month shelf life, room-temperature storage (18 - 28°C / 65 -82 °F)
  • Compatible with standard capillary collection tubes, supporting flexible downstream processing
  • Fits seamlessly into kitted solutions with optional fulfillment, tracking, and end-to-end logistics support
Specifications
Color White
Additive None
Material Plastic
Sterility Non-sterile
Closure Type Cap
Capacity (Metric) 500 μL