Learn More
Grainger Huot Tap and Drill Index, NC and NF, Steel, 36 Compartments, 1 ea
Catalog No. 502788141
Encompass_Preferred
This heavy-gauge steel tap and drill index organizes National Coarse (UNC) and National Fine (UNF) tools in a single, durable storage unit. Designed for efficient shop use, the index provides 36 compartments that hold taps and their corresponding drill bits side by side for quick identification. The case features a baked-on epoxy finish for corrosion resistance and a secure, snap-lock cover to keep tools organized and protected.
  • Holds up to 36 taps and drill bits for both UNC and UNF sizes.
  • Features raised, embossed lettering for permanent tool identification.
  • Includes internal clips in the cover for securing a T-handle or adjustable tap wrench.
  • Constructed from rugged, heavy-gauge steel with a protective epoxy finish.
  • Provides precise storage holes for quick tool selection.
  • Note: Taps and drills are sold separately.
Catalog No. 50-278-8141 Supplier Grainger Supplier No. 18EC40
This heavy-gauge steel tap and drill index organizes National Coarse (UNC) and National Fine (UNF) tools in a single, durable storage unit. Designed for efficient shop use, the index provides 36 compartments that hold taps and their corresponding drill bits side by side for quick identification. The case features a baked-on epoxy finish for corrosion resistance and a secure, snap-lock cover to keep tools organized and protected.
  • Holds up to 36 taps and drill bits for both UNC and UNF sizes.
  • Features raised, embossed lettering for permanent tool identification.
  • Includes internal clips in the cover for securing a T-handle or adjustable tap wrench.
  • Constructed from rugged, heavy-gauge steel with a protective epoxy finish.
  • Provides precise storage holes for quick tool selection.
  • Note: Taps and drills are sold separately.