Learn More
Grainger Huot Tap and Drill Index, Steel, 18 Compartments, 1 ea
Catalog No. 502784620
Encompass_Preferred
This tap and drill index provides an organized, rugged storage solution for National Coarse (NC) tools. Constructed from heavy gauge steel, this unit features 18 precisely sized compartments to ensure quick tool selection and storage for taps ranging from 6-32 to 1/2 in.-13 and their corresponding drills.
  • Constructed from heavy gauge all-steel for durability
  • Features 18 compartments for organized tool storage
  • Embossed lettering for permanent identification
  • Finished with a corrosion-resistant, epoxy baked-on coating
  • Equipped with a secure, snap-lock steel cover
  • Compact dimensions of 10.16 cm (4 in.) W x 17.78 cm (7 in.) D x 3.18 cm (1.25 in.) H
Catalog No. 50-278-4620 Supplier Grainger Supplier No. 18EC37
This tap and drill index provides an organized, rugged storage solution for National Coarse (NC) tools. Constructed from heavy gauge steel, this unit features 18 precisely sized compartments to ensure quick tool selection and storage for taps ranging from 6-32 to 1/2 in.-13 and their corresponding drills.
  • Constructed from heavy gauge all-steel for durability
  • Features 18 compartments for organized tool storage
  • Embossed lettering for permanent identification
  • Finished with a corrosion-resistant, epoxy baked-on coating
  • Equipped with a secure, snap-lock steel cover
  • Compact dimensions of 10.16 cm (4 in.) W x 17.78 cm (7 in.) D x 3.18 cm (1.25 in.) H