Learn More
Grainger TE-CO T-Slot Nut, Low Carbon Steel, 3/8-16 Thread, 25/Pk.
Catalog No. 502789802
Encompass_Preferred
This tap-through T-slot nut is designed for secure fastening applications within machine table slots. Engineered from grade 1018 low carbon steel with a black oxide finish, it provides reliable durability for industrial workholding needs.
  • Features a 3/8 in.-16 UNC coarse thread
  • Designed with a tap-through configuration for versatile installation
  • Constructed from case-hardened, low carbon steel for strength
  • Finished with black oxide for corrosion resistance
  • Compatible with 1/2 in. slot sizes and 13 mm table slots
  • Made in the USA and TAA compliant
Catalog No. 50-278-9802 Supplier Grainger Supplier No. 169P92
This tap-through T-slot nut is designed for secure fastening applications within machine table slots. Engineered from grade 1018 low carbon steel with a black oxide finish, it provides reliable durability for industrial workholding needs.
  • Features a 3/8 in.-16 UNC coarse thread
  • Designed with a tap-through configuration for versatile installation
  • Constructed from case-hardened, low carbon steel for strength
  • Finished with black oxide for corrosion resistance
  • Compatible with 1/2 in. slot sizes and 13 mm table slots
  • Made in the USA and TAA compliant