Learn More
Grainger TE-CO T-Slot Nut, 1018 Steel, 3/4 in Slot, 5/8 in-11 Thread, 5/Pk.
Catalog No. 502788395
Encompass_Preferred
This tapped-through T-slot nut is designed for secure mounting in 3/4 in. (20 mm) machine table slots. Constructed from 1018 steel with a black oxide finish, it provides reliable durability and wear resistance in demanding machining environments. The nut features a fully threaded design, allowing the stud to pass completely through for maximum versatility in fixture setup.
  • Constructed from 1018 steel with a protective black oxide finish
  • Designed for 3/4 in. (20 mm) machine table slots
  • 5/8-11 UNC thread size
  • Fully threaded design allows stud to pass through the base
  • Conforms to TCMAI standards
Catalog No. 50-278-8395 Supplier Grainger Supplier No. 169P11
This tapped-through T-slot nut is designed for secure mounting in 3/4 in. (20 mm) machine table slots. Constructed from 1018 steel with a black oxide finish, it provides reliable durability and wear resistance in demanding machining environments. The nut features a fully threaded design, allowing the stud to pass completely through for maximum versatility in fixture setup.
  • Constructed from 1018 steel with a protective black oxide finish
  • Designed for 3/4 in. (20 mm) machine table slots
  • 5/8-11 UNC thread size
  • Fully threaded design allows stud to pass through the base
  • Conforms to TCMAI standards