Learn More
Aviva Systems Biology Corp TAP1 Antibody - middle region (ARP43628_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43628_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TAP1. Alternate names: ABC17, ABCB2, APT1, D6S114E, FLJ26666, FLJ41500, PSF1, RING4, TAP1*0102N, TAP1N, PSF-1ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58592