Learn More
Cayman Chemical TAPA1CD81 EXrAClular Dom 100ug
SDP
Catalog No. 502919365
Encompass_Preferred
Recombinant N-terminal human His-tagged CD81 expressed in HEK293 cells • Amino acids: 113-201 • MW: 12.1 kDa
Catalog No. 50-291-9365 Supplier Cayman Chemical Supplier No. 37019100 UG
Recombinant N-terminal human His-tagged CD81 expressed in HEK293 cells • Amino acids: 113-201 • MW: 12.1 kDa