Learn More
Aviva Systems Biology Corp TAPBP Antibody - C-terminal region (ARP45437_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45437_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAPBP. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TAPBP. Alternate names: NGS17, TAPA, TPN, TPSNApplicationWB, IHC. Species reactivity: Cow, Human, Pig, Sheep
Catalog No. 50-014-59028