Learn More
Aviva Systems Biology Corp TAPBP Antibody - N-terminal region (ARP45436_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45436_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TAPBP. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human TAPBP. Alternate names: NGS17, TAPA, TPN, TPSNApplicationWB, IHC. Species reactivity: Dog, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-285-199